Learn More
Newcomer Supply Giemsa Stock Stain, Wolbach, 500 ml
SDP
Catalog No. NC9527279
Encompass
Use: Wolbach & May-Grunwald mod. Giemsa Stain. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9527279 Supplier Newcomer Supply Supplier No. 1121A
Use: Wolbach & May-Grunwald mod. Giemsa Stain. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.