Learn More
Electron Microscopy Sciences Giemsa Stock Solution 250 ML
SDP
Catalog No. 5031701
Encompass_Preferred
Used in Giemsa Thick Film Stain, Lillie (1965); Barber and Komp (1929. Stain malarial parasites. For references see Clark, G. Staining Procedures, Williams and Wilkins Company, Baltimore, 3rd Ed., p. 126, c. 1973 and Lillie, R.D. Histologic Technic and Practical Histochemistry, 3rd Ed., McGraw-Hill, New York, 1965.
Catalog No. 50-317-01 Supplier Electron Microscopy Sciences Supplier No. 2615403
Used in Giemsa Thick Film Stain, Lillie (1965); Barber and Komp (1929. Stain malarial parasites. For references see Clark, G. Staining Procedures, Williams and Wilkins Company, Baltimore, 3rd Ed., p. 126, c. 1973 and Lillie, R.D. Histologic Technic and Practical Histochemistry, 3rd Ed., McGraw-Hill, New York, 1965.