Learn More
StatLab Giemsa/Wright Control Slides
Description
- Standardize and satisfy regulatory requirements for Giemsa-Wright Stain controls and use with malaria parasite staining procedures
- Scan quickly with high organism counts of red blood cells infected with Babesia sp.
- Eliminates the need to maintain your own cultures and are cost effective compared to alternative brands
- 10 slides/box
Specifications
Specifications
| Product Type | Control Slide |
| Quantity | 10 Slides/Box |