Learn More
Agilent Technologies GIGAPACK III XL PACK EXTRACT
Catalog No. 50125038
Encompass
Gigapack III XL Packaging Extract 11 vials Packaging efficiency: 1 x 109 pfu/microgram. Preferentially packages phage with large inserts; For genomic lambda and cosmid libraries Contains: Gigapack III XL packaging extract
Catalog No. 50-125-038 Supplier Agilent Technologies Supplier No. 200209
Gigapack III XL Packaging Extract 11 vials Packaging efficiency: 1 x 109 pfu/microgram. Preferentially packages phage with large inserts; For genomic lambda and cosmid libraries Contains: Gigapack III XL packaging extract