Learn More
Agilent Technologies GIGAPACK III XL PACK EXTRACT
Supplier:  Agilent Technologies 200207
Encompass
Gigapack III XL packaging extract is used to package recombinant lambda phage with selectivity for phage containing large inserts. Preferentially packages phage with large inserts; For genomic lambda and cosmid libraries
Catalog No. 50-125-036