Learn More
Surgical Design™ Gigli Wire Saw System (Handle For Wire Saw)
Catalog No. 21124653 Shop All Surgical Design Products
Change view
Click to view available options
Shape:
Loop Grip
Snap Lock
Standard
T Shape, Turnable
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape
21-124-653 Snap Lock
21-124-640 T Shape, Turnable
21-124-641 Loop Grip
21-124-652 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-124-653 Supplier Surgical Design™ Supplier No. ATPM032
Add to Cart
Add to Cart
Gigli wire saw system delivers controlled bone cutting for cranial, thoracic, and pelvic postmortem procedures.
  • Complete Gigli Wire Saw System designed for autopsy and postmortem bone cutting
  • Includes Gigli Wire Saw Blades (available in 12 in., 16 in., and 20 in.) and multiple compatible handle styles
  • Two handles are required for proper bilateral traction when using any Gigli wire blade
  • Flexible serrated wire blades allow controlled cutting in tight or curved anatomical regions
  • Ideal for skullcap removal, thoracic entry, rib separation, pelvic cutting, and long-bone sectioning
  • Produces a smooth, even cutting motion with reduced debris—important for forensic clarity
  • Made from high-tensile stainless steel for durability, bend resistance, and repeated autoclaving
  • Compatible handle styles include:Gigli Strully T-Handle Modified Turnable (reduces wire twisting during long cuts)
  • Gigli Strully T-Handle Modified Loop Grip (improved traction with gloved hands)
  • Standard Gigli T-Handle (classic solid design for consistent control)
  • Rigid T-Shape Snap-Lock Handle (secure locking mechanism for fast wire attachment)
  • Handles constructed from surgical-grade stainless steel, corrosion-resistant, and fully autoclavable
  • Ergonomic gripping surfaces reduce operator fatigue during high-force bone work
  • Designed for forensic pathology, hospital autopsy, medical examiners, and necropsy labs
  • Cranial autopsy (skull cutting around curved calvarium)
  • Opening the thoracic cavity by cutting ribs
  • Sectioning pelvic, vertebral, or long bones
  • Ideal for confined anatomical locations where rigid saws cannot operate
  • Suitable for both human autopsy and veterinary necropsy
Specifications
Product Type Handle, Snap-Lock
Certifications/Compliance ISO/GMP
Material Stainless Steel
Shape Snap Lock
Sterility Non-sterile