Learn More
StatLab™ Gill 1 Hematoxylin Stain
GSA_VASDP
Catalog No. STLSL971
Change view
Click to view available options
Quantity:
4 x 1 gal.
4 x 16 oz. Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSL971 4 x 1 gal.
STLSL9716 4 x 16 oz. Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL971 Supplier StatLab™ Supplier No. SL971
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Low dye concentration for cytology.
  • Used progressively, this formulation optimally stains both gynecological and non-gynecological specimens
  • Produces high nuclear clarity
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 1 gal.
Product Type Hematoxylin