Learn More
StatLab™ Gill 2 Hematoxylin Stain
GSA_VASDP
Catalog No. STLSL9416
Change view
Click to view available options
Quantity:
4 x 1 gal.
4 x 16 oz. Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSL9416 4 x 16 oz. Bottle
STLSL941 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL9416 Supplier StatLab™ Supplier No. SL9416
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Medium dye concentration for histology or intense cytology staining.
  • Produces a darker nuclei, or used when shorter staining times are needed
  • Nuclear detail is very distinct and has great clarity
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Application) Histology or Cytology Staining
Quantity 4 x 16 oz. Bottle
Product Type Hematoxylin