Learn More
Epredia™ Gill™ Hematoxylin 1, 2, and 3
Catalog No. 22050199 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
Product Type:
Gill 1 Hematoxylin
Gill 2 Hematoxylin
Gill 3 Hematoxylin
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-199 1 pt. Gill 1 Hematoxylin
22-050-200 1 gal. Gill 1 Hematoxylin
22-050-201 1 pt. Gill 2 Hematoxylin
22-050-202 1 gal. Gill 2 Hematoxylin
22-050-203 1 pt. Gill 3 Hematoxylin
22-050-204 1 gal. Gill 3 Hematoxylin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-050-199 Supplier Epredia™ Supplier No. 72404
Add to Cart
Add to Cart
With three strengths available, the Epredia™ Gill™ 1, 2, 3 Hematoxylin allows complete control of staining intensities.
Gill Hematoxylins are progressive nuclear stains.
  • Gill 1 is recommended for use in cytology
  • Gill 2 has applications in histology and cytology
  • Gill 3 is used in histology
  • All are pre-mixed and ready-to-use
Specifications
Quantity 1 pt.
Product Type Gill 1 Hematoxylin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov