Description
- Gill 1 is recommended for use in cytology
- Gill 2 has applications in histology and cytology
- Gill 3 is used in histology
- All are pre-mixed and ready-to-use
Specifications
Specifications
| Quantity | 1 gal. |
| Product Type | Gill 1 Hematoxylin |