Learn More
Leica Biosystems GILL I HEMATOXYLIN 1 GAL
Catalog No. NC2056556
Encompass
Three Gill formulations are available: The Gill I formula is generally recommended for cytology. The stronger Gill III is usually preferred for use in histology. GIll II is a medium strength hematoxylin. The Gill formula hematoxylins are used in the progressive method of staining. The Gill formulations do not contain mercury. Available in various quantities.
This item is not returnable. View return policy
Catalog No. NC2056556 Supplier Leica Biosystems Supplier No. 3801500
Three Gill formulations are available: The Gill I formula is generally recommended for cytology. The stronger Gill III is usually preferred for use in histology. GIll II is a medium strength hematoxylin. The Gill formula hematoxylins are used in the progressive method of staining. The Gill formulations do not contain mercury. Available in various quantities.