Learn More
Leica Biosystems GILL II HEMATOXYLIN (GAL)
Supplier:  Leica Biosystems 3801520
Encompass_Preferred
Three Gill formulations are available:The Gill I formula is generally recommended for cytology.The stronger Gill III is usually preferred for use in histology.GIll II is a medium strength hematoxylin.The Gill formula hematoxylins are used in the progressive method of staining.The Gill formulations do not contain mercury.Available in various quantities.
Catalog No. 50-255-2441