Learn More
Electron Microscopy Sciences Gill's Hematoxylin No. 1, 1L
SDP
Catalog No. 5026110
Encompass_Preferred
Gill's Hematoxylin is a specially formulated solution for use as a biological stain for the nuclei and nuclear chromatin of normal and abnormal cells whether in whole or sliced cells. Gill's Hematoxylin is designed to use at its full strength. It is microfiltered prior to shipment; therefore, filtration before use is not required. For repeated use further filtration is recommended.
Catalog No. 50-261-10 Supplier Electron Microscopy Sciences Supplier No. 2603010
Gill's Hematoxylin is a specially formulated solution for use as a biological stain for the nuclei and nuclear chromatin of normal and abnormal cells whether in whole or sliced cells. Gill's Hematoxylin is designed to use at its full strength. It is microfiltered prior to shipment; therefore, filtration before use is not required. For repeated use further filtration is recommended.