Learn More
Electron Microscopy Sciences Gill's Hematoxylin #2 4 LT
SDP
Catalog No. 5026111
Encompass_Preferred
Gill's Hematoxylin is a specially formulated solution for use as a biological stain for the nuclei and nuclear chromatin of normal and abnormal cells whether in whole or sliced cells.
Our Gill's Hematoxylin is designed to use at its full strength. It is microfiltered prior to shipment; therefore, filtration before use is not required. For repeated use further filtration is recommended. May be diluted with ethylene glycol (25% is recommended).
Catalog No. 50-261-11 Supplier Electron Microscopy Sciences Supplier No. 2603024
Gill's Hematoxylin is a specially formulated solution for use as a biological stain for the nuclei and nuclear chromatin of normal and abnormal cells whether in whole or sliced cells.
Our Gill's Hematoxylin is designed to use at its full strength. It is microfiltered prior to shipment; therefore, filtration before use is not required. For repeated use further filtration is recommended. May be diluted with ethylene glycol (25% is recommended).