Learn More
Polysciences, Inc. Gill's Hematoxylin #1; Polysciences; For cytology; Composition: acetic ccid; aluminum sulfate; ethylene glycol, hematoxylin, sodium iodate, water 500mL
SDP
Catalog No. NC9168673
Encompass_Preferred
Gill's Hematoxylin #1; Polysciences; For cytology; Composition: acetic ccid; aluminum sulfate; ethylene glycol, hematoxylin, sodium iodate, water; Store at 25 deg.C, SG:1.03; Stains gynecological, non-gynecological specimens; 500mL
Catalog No. NC9168673 Supplier Polysciences, Inc. Supplier No. 24242500
Gill's Hematoxylin #1; Polysciences; For cytology; Composition: acetic ccid; aluminum sulfate; ethylene glycol, hematoxylin, sodium iodate, water; Store at 25 deg.C, SG:1.03; Stains gynecological, non-gynecological specimens; 500mL