Learn More
Polysciences, Inc. Gills Hematoxylin #2, double strength for Histology & Cytology, 1412-19-7, MFCD00078111, 500 mL
SDP
Catalog No. NC9480878
Encompass_Preferred
HEMATOXYLIN, Molecular Formula: C16 H14 O6, Molecular Weight: 302.281. Polysciences, Inc. Gold Standard Series Hematoxylin and Eosin products offer you the highest quality for routine staining. Strict quality control for all Gold Standard Series Routine Stains ensures lot-to-lot consistency. Stains are packaged in convenient plastic recyclable containers. General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining. Storage: Store at room temperature.
Catalog No. NC9480878 Supplier Polysciences, Inc. Supplier No. 24243500
HEMATOXYLIN, Molecular Formula: C16 H14 O6, Molecular Weight: 302.281. Polysciences, Inc. Gold Standard Series Hematoxylin and Eosin products offer you the highest quality for routine staining. Strict quality control for all Gold Standard Series Routine Stains ensures lot-to-lot consistency. Stains are packaged in convenient plastic recyclable containers. General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining. Storage: Store at room temperature.