Learn More
Polysciences, Inc. Gills modified OG-6, 1000 mL
SDP
Catalog No. NC9002566
Encompass
OG-6 is a cytoplasmic counterstain solution used in sequence with EA (EA 50, EA 65 or EA36) in the Papanicolaou staining method for clinical cytology. Gills modified OG-6 is stable in solution and gives predictable high quality staining results not previously possible with other formulations. Hazards: Flammable liquid, Handling: Gloves & fume hood, Storage: Store at room temperature.
Catalog No. NC9002566 Supplier Polysciences, Inc. Supplier No. 097821000
OG-6 is a cytoplasmic counterstain solution used in sequence with EA (EA 50, EA 65 or EA36) in the Papanicolaou staining method for clinical cytology. Gills modified OG-6 is stable in solution and gives predictable high quality staining results not previously possible with other formulations. Hazards: Flammable liquid, Handling: Gloves & fume hood, Storage: Store at room temperature.