Learn More
Med Vet International Gillies Retractor Fine, 7in, J0243G, Each
Catalog No. 5000585871
Encompass_Preferred
Very delicate hook tip for retraction of fine nerves and vessels. Stainless steel.
Catalog No. 50-005-85871 Supplier Med Vet International Supplier No. JOR243G
Very delicate hook tip for retraction of fine nerves and vessels. Stainless steel.