Learn More
Electron Microscopy Sciences Gills Hematoxylin #1 500 ML
SDP
Catalog No. 5030119
Encompass_Preferred
Used in Papanicolaou Stain for Membrane Filter. Reference: Gill, G.W., ActaCytol., 18:300-311, 1974, Diagnostic Cytology by Membrane Filter, Application Bulletin 100, Gelman Instrument Co., Ann Arbor, Michigan, 1978
Catalog No. 50-301-19 Supplier Electron Microscopy Sciences Supplier No. 2680101
Used in Papanicolaou Stain for Membrane Filter. Reference: Gill, G.W., ActaCytol., 18:300-311, 1974, Diagnostic Cytology by Membrane Filter, Application Bulletin 100, Gelman Instrument Co., Ann Arbor, Michigan, 1978