Learn More
Gilson Company Inc 3" Sieve, Brass/Stainless, Full Height, No. 80
Catalog No. NC0203652
Encompass_Preferred
3in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 80 (180µm) openings and full height brass frame. The overall height is 1-3/4in and the stacked height is 1-1/8in.
Catalog No. NC0203652 Supplier Gilson Company Inc Supplier No. V3CF #80
3in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 80 (180µm) openings and full height brass frame. The overall height is 1-3/4in and the stacked height is 1-1/8in.