Learn More
Gilson Company Inc No. 10 Mesh Sieve, All Stainless, Full Height, 8" diameter
Catalog No. NC1838417
Encompass_Preferred
8in (203mm) diameter ASTM E11 Test Sieve is built with No. 10 (2.0mm) stainless steel woven wire cloth and a full-height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in.
Catalog No. NC1838417 Supplier Gilson Company Inc Supplier No. V8SF#10
8in (203mm) diameter ASTM E11 Test Sieve is built with No. 10 (2.0mm) stainless steel woven wire cloth and a full-height stainless steel frame. The overall height is 2-5/8in and the stacked height is 2-1/8in.