Learn More
Gilson Company Inc Indicating Drierite, No. 8 Granules, 5 lb jar
Catalog No. NC1301046
Encompass
Indicating Drierite is used to rapidly absorb water vapor from air and gases. Made from cobalt chloride impregnated granules, the blue Drierite crystals change to a distinct pink color when exhausted. Indicating Drierite can be regenerated in an oven for longer life or can be replaced as needed.
Catalog No. NC1301046 Supplier Gilson Company Inc Supplier No. SGA106
Indicating Drierite is used to rapidly absorb water vapor from air and gases. Made from cobalt chloride impregnated granules, the blue Drierite crystals change to a distinct pink color when exhausted. Indicating Drierite can be regenerated in an oven for longer life or can be replaced as needed.