Learn More
Pipette Supplies, Inc. Gilson Pipetman Classic / Pipetman Neo Friction Ring, Newer Version (Pipette Supplies)
SDP
Catalog No. NC1908812
Encompass
The friction ring for the Gilson Pipetman Classic and Newer Style Gilson Pipetman Neo. These friction rings (tension rings) are thinner and fit over the hard plastic band on the newer (post Feb 1991) style Gilson Pipetman.Made in the USA
Catalog No. NC1908812 Supplier Pipette Supplies, Inc. Supplier No. 40000/5
The friction ring for the Gilson Pipetman Classic and Newer Style Gilson Pipetman Neo. These friction rings (tension rings) are thinner and fit over the hard plastic band on the newer (post Feb 1991) style Gilson Pipetman.Made in the USA