Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMANTM DiamondTM Tips Blister Refill
GREENER_CHOICE

Catalog No. F172401G Shop All Gilson Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Not Autoclavable
Sterility:
Non-sterile
Sterile
Model:
D300; Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable Sterility Model
F172401G Not Autoclavable Sterile D300; Standard
F172400G Autoclavable Non-sterile D300; Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F172401G Supplier Gilson™ Supplier No. F172401
Only null left
Add to Cart
Add to Cart

Individually wrapped racks of tips that are convenient, clean and eco-friendly

  • Color-coded racks to indicate tip models and PIPETMAN model compatibility
  • Available as autoclavable or gamma-ray sterilized
  • Fit with PIPETMAX (special tips validated for use with 384 well plates)
  • 10 individually wrapped racks of 96 tips
  • Ready to use

Specifications

Volume (Metric) 20 to 300 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model D300; Standard
Length (English) 10.87 in.
Length (Metric) 5.1cm
Clean Claims DNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free
Format Blister Refill
Tip Style Standard
Quantity 960 (10x96)
Product Type Air Displacement Tips
Disposable Single-use
For Use With (Equipment) Pipetmax
Graduated No
Product Line Pipetman Diamond
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.