Learn More
Pipette Supplies, Inc. Gilson Pipetman G & L O-ring, Inner, P1000G, P1000L (Pipette Supplies)
SDP
Catalog No. NC1908819
Encompass
Replacement o-ring for:Gilson Pipetman G 1000μL (P1000G)Gilson Pipetman L 1000μL (P1000L)Smaller / thinner of the two o-rings for this pipette. The smaller seals to the piston; the larger seals to the tip holder. There is a spacer in between.Made in the USA
Catalog No. NC1908819 Supplier Pipette Supplies, Inc. Supplier No. 3L1001/5
Replacement o-ring for:Gilson Pipetman G 1000μL (P1000G)Gilson Pipetman L 1000μL (P1000L)Smaller / thinner of the two o-rings for this pipette. The smaller seals to the piston; the larger seals to the tip holder. There is a spacer in between.Made in the USA