Learn More
Gilson Company Inc GILSON SPINNING RIFFLER MODEL
Catalog No. NC2306481
Encompass_Preferred
  • Exceptional accuracy for reliable dry material division.
  • Intuitive touchscreen control for precise adjustment of rotation speed and vibration amplitude.
  • Consistent material feed with a vibratory feeder and a 1 L stainless steel hopper.
  • Enhanced safety and quiet operation with isolated motor and enclosed drum.
  • Versatile sample collection into sixteen 60 ml vessels or threaded vials.
  • Durable construction using urethane-based resin and stainless steel.
  • Energy-efficient 'sleep' mode for power conservation.
  • Meets ASTM D2013 and ASTM B215 standards for material sampling.
  • Dimensions: 13 in x 20.5 in x 23 in.
  • Estimated shipping weight: 45 lb.
Catalog No. NC2306481 Supplier Gilson Company Inc Supplier No. SP230
Add to Cart
Add to Cart
Request a Quote
  • Exceptional accuracy for reliable dry material division.
  • Intuitive touchscreen control for precise adjustment of rotation speed and vibration amplitude.
  • Consistent material feed with a vibratory feeder and a 1 L stainless steel hopper.
  • Enhanced safety and quiet operation with isolated motor and enclosed drum.
  • Versatile sample collection into sixteen 60 ml vessels or threaded vials.
  • Durable construction using urethane-based resin and stainless steel.
  • Energy-efficient 'sleep' mode for power conservation.
  • Meets ASTM D2013 and ASTM B215 standards for material sampling.
  • Dimensions: 13 in x 20.5 in x 23 in.
  • Estimated shipping weight: 45 lb.