Learn More
Medchemexpress LLC HY-12432 100mg , Gilteritinib CAS:1254053-43-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12432100MG
Encompass_Preferred
HY-12432 100mg Gilteritinib CAS: 1254053-43-4 Gilteritinib (ASP2215) is a potent and ATP-competitive FLT3/AXL inhibitor with IC50s of 0.29 nM/0.73 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9043