Learn More
Cayman Chemical Gilteritinib, Purity: ≥98%, CAS: 1254053-43-4, 10 mg
SDP
Catalog No. 502987481
Encompass_Preferred
A FLT3 inhibitor (IC50 = 5 nM for the wild-type enzyme); inhibits various FLT3 mutants (IC50s = 1.4-12.2 nM); also inhibits Axl and c-Kit (IC50s = 41 and 102 nM, respectively); decreases the viability of blast cells isolated from patients with relapsed AML in a concentration-dependent manner
Catalog No. 50-298-7481 Supplier Cayman Chemical Supplier No. 2150310 MG
A FLT3 inhibitor (IC50 = 5 nM for the wild-type enzyme); inhibits various FLT3 mutants (IC50s = 1.4-12.2 nM); also inhibits Axl and c-Kit (IC50s = 41 and 102 nM, respectively); decreases the viability of blast cells isolated from patients with relapsed AML in a concentration-dependent manner