Learn More
Medchemexpress LLC HY-12432 10mg Medchemexpress, Gilteritinib CAS:1254053-43-4 Purity:>98%
SDP
Catalog No. 502028878
Encompass_Preferred
Medchemexpress, HY-12432 10mg Gilteritinib CAS:1254053-43-4 Gilteritinib is a potent FLT3/AXL inhibitor with IC50s of 0.29 nM/0.73 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8878 Supplier Medchemexpress LLC Supplier No. HY1243210MG
Medchemexpress, HY-12432 10mg Gilteritinib CAS:1254053-43-4 Gilteritinib is a potent FLT3/AXL inhibitor with IC50s of 0.29 nM/0.73 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.