Learn More
Medchemexpress LLC Gilteritinib | 1254053-43-4 | 200MG
SDP
Catalog No. 5000235401
Encompass_Preferred
Gilteritinib (ASP2215) is a potent and ATP-competitive FLT3/AXL inhibitor with IC50s of 0 29 nM/0 73 nM respectively
Catalog No. 50-002-35401 Supplier Medchemexpress LLC Supplier No. HY12432200MG
Gilteritinib (ASP2215) is a potent and ATP-competitive FLT3/AXL inhibitor with IC50s of 0 29 nM/0 73 nM respectively