Learn More
Medchemexpress LLC Gilteritinib | 1254053-43-4 | MFCD28144685 | 99.6% | 552.71 g·mol⁻¹ | C29H44N8O3 | 10MG
SDP
Catalog No. 5000238652
Encompass_Preferred
Gilteritinib analytical standard provided as a high-purity reference material for research and analytical use. It is intended for method development, instrument calibration, and quantitative assays that require a reliable reference substance of the FLT3/AXL inhibitor gilteritinib.
Catalog No. 50-002-38652 Supplier Medchemexpress LLC Supplier No. HY12432R10MG
Gilteritinib analytical standard provided as a high-purity reference material for research and analytical use. It is intended for method development, instrument calibration, and quantitative assays that require a reliable reference substance of the FLT3/AXL inhibitor gilteritinib.