Learn More
Selleck Chemical LLC Gilteritinib
SDP
Supplier:  Selleck Chemical LLC S775425MG
Encompass_Preferred
Gilteritinib (ASP2215) is a small-molecule FLT3/AXL inhibitor with IC50 values of 0 29 nM and 0 73 nM for FLT3 and AXL respectively It inhibits FLT3 at an IC50 value that was approximately 800-fold more potent than the concentration required to inhibit c-KIT (230 nM)
Catalog No. 50-000-00601