Learn More
Selleck Chemical LLC Gilteritinib
SDP
Catalog No. 5000001355
Encompass_Preferred
Gilteritinib (ASP2215) is a small-molecule FLT3/AXL inhibitor with IC50 values of 0 29 nM and 0 73 nM for FLT3 and AXL respectively It inhibits FLT3 at an IC50 value that was approximately 800-fold more potent than the concentration required to inhibit c-KIT (230 nM)
Catalog No. 50-000-01355 Supplier Selleck Chemical LLC Supplier No. S7754100MG
Gilteritinib (ASP2215) is a small-molecule FLT3/AXL inhibitor with IC50 values of 0 29 nM and 0 73 nM for FLT3 and AXL respectively It inhibits FLT3 at an IC50 value that was approximately 800-fold more potent than the concentration required to inhibit c-KIT (230 nM)