Learn More
Selleck Chemical LLC Gilteritinib (ASP2215) 5mg 1254053-43-4
SDP
Supplier:  Selleck Chemical LLC S77545MG
Encompass_Preferred
Gilteritinib (ASP2215) is a small-molecule FLT3/AXL inhibitor with IC50 values of 0.29 nM and 0.73 nM for FLT3 and AXL, respectively. It inhibits FLT3 at an IC50 value that was approximately 800-fold more potent than the concentration required to inhibit c-KIT (230 nM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8003