Learn More
TARGETMOL CHEMICALS INC Gilteritinib hemifumarate 10MG
SDP
Catalog No. 502877857
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Gilteritinib hemifumarate (ASP2215 hemifumarate) is a potent ATP-competitive dual FLT3 (IC50 0.29 nM) and AXL (IC50 0.73 nM) inhibitor for the treatment of relapsed or refractory FLT3 mutant AML. Purity 98.07%
Catalog No. 50-287-7857 Supplier TARGETMOL CHEMICALS INC Supplier No. T7197310MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Gilteritinib hemifumarate (ASP2215 hemifumarate) is a potent ATP-competitive dual FLT3 (IC50 0.29 nM) and AXL (IC50 0.73 nM) inhibitor for the treatment of relapsed or refractory FLT3 mutant AML. Purity 98.07%