Learn More
Cayman Chemical GIlvocarcIn M 2.5mg
SDP
Catalog No. 502935862
Encompass_Preferred
An antibiotic; active against S. aureus bacteria at 32 UG/ml; inhibits growth of KB cells (IC50 = 0.52 UG/ml); intercalates into bacteriophage PM2 DNA; toxic to rats (LD50 = 450 mg/kg, i.v.)
Catalog No. 50-293-5862 Supplier Cayman Chemical Supplier No. 257622.5 MG
An antibiotic; active against S. aureus bacteria at 32 UG/ml; inhibits growth of KB cells (IC50 = 0.52 UG/ml); intercalates into bacteriophage PM2 DNA; toxic to rats (LD50 = 450 mg/kg, i.v.)