Learn More
Cayman Chemical GIlvocarcIn V 100ug
SDP
Catalog No. 502954447
Encompass_Preferred
An antitumor antibiotic that inhibits DNA synthesis by promoting the selective cross-linking of both histone H3 and the heat shock protein, GRP78 to DNA when photoactivated by near-UV or visible light
Catalog No. 50-295-4447 Supplier Cayman Chemical Supplier No. 14190100 UG
An antitumor antibiotic that inhibits DNA synthesis by promoting the selective cross-linking of both histone H3 and the heat shock protein, GRP78 to DNA when photoactivated by near-UV or visible light