Learn More
AdipoGen Gilvocarcin V
SDP
Catalog No. 502562035
Encompass_Preferred
Chemical. CAS 77879-90-4. Formula C27H26O9. MW 494.5. Isolated from Streptomyces sp. Gö 3592. Antibiotic. Antitumor compound. Weakly active against Gram-positive bacteria and fungi. Mediates a unique cross-linking reaction between DNA and histone H3 by light. Single strand scission and covalent binding to DNA after photoactivation.
Catalog No. 50-256-2035 Supplier AdipoGen Supplier No. BVT0256C250
Add to Cart
Add to Cart
Chemical. CAS 77879-90-4. Formula C27H26O9. MW 494.5. Isolated from Streptomyces sp. Gö 3592. Antibiotic. Antitumor compound. Weakly active against Gram-positive bacteria and fungi. Mediates a unique cross-linking reaction between DNA and histone H3 by light. Single strand scission and covalent binding to DNA after photoactivation.