Learn More
Aviva Systems Biology Corp GIMAP5 Antibody - middle region (ARP47440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GIMAP5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GIMAP5. Alternate names: FLJ11296, HIMAP3, IAN4, IAN4L1, IAN5, IMAP3, hIAN5, IROD, IAN-5ApplicationWB. Species reactivity: Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59552