Learn More
Aviva Systems Biology Corp GIMAP6 Antibody - C-terminal region (ARP56178_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56178_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GIMAP6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GIMAP6. Alternate names: DKFZp686A01175, FLJ22690, IAN6, hIAN2, IAN2, IAN-2, IAN-6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-122