Learn More
Medchemexpress LLC HY-B0063 5mg Medchemexpress, Gimatecan CAS:292618-32-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB00635MG
Encompass_Preferred
Medchemexpress, HY-B0063 5mg Gimatecan CAS:292618-32-7 Gimatecan (ST1481) is a potent topoisomerase I inhibitor. Gimatecan is an orally bioavailable camptothecin analogue with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2351