Learn More
Medchemexpress LLC HY-17469 25mg Medchemexpress, Gimeracil CAS:103766-25-2 Purity:>98%
SDP
Catalog No. 502081960
Encompass_Preferred
Medchemexpress, HY-17469 25mg Gimeracil CAS:103766-25-2 Gimeracil, a component of an oral fluoropyrimidine derivative S-1, inhibits DNA DSB repair and is a potent inhibitor of DPYD (dihydropyrimidine dehydrogenase, DPD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1960 Supplier Medchemexpress LLC Supplier No. HY1746925MG
Medchemexpress, HY-17469 25mg Gimeracil CAS:103766-25-2 Gimeracil, a component of an oral fluoropyrimidine derivative S-1, inhibits DNA DSB repair and is a potent inhibitor of DPYD (dihydropyrimidine dehydrogenase, DPD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.