Learn More
Selleck Chemical LLC Tetrahydropalmatine hydrochloride S3854-25mg
SDP
Catalog No. 503108311
Encompass_Preferred
Tetrahydropalmatine (THP Gindarine 1-Tetrahydropalmitine) is an isoquinoline alkaloid found in several different plant species mainly in the Corydalis genus (Yan Hu Suo) but also in other plants such as Stephania rotunda It is a potent muscle relaxant Tetrahydropalmatine hydrochloride acts through inhibition of amygdaloid release of dopamine to inhibit an epileptic attack in rats
Catalog No. 50-310-8311 Supplier Selleck Chemical LLC Supplier No. S385425MG
Tetrahydropalmatine (THP Gindarine 1-Tetrahydropalmitine) is an isoquinoline alkaloid found in several different plant species mainly in the Corydalis genus (Yan Hu Suo) but also in other plants such as Stephania rotunda It is a potent muscle relaxant Tetrahydropalmatine hydrochloride acts through inhibition of amygdaloid release of dopamine to inhibit an epileptic attack in rats