Learn More
Enzo Life Sciences Gingerol (20mg). CAS: 23513-14-6
SDP
Supplier:  Enzo Life Sciences BMLCA4220020
Encompass_Preferred
Gingerol ([6]-gingerol), a pungent vanilloid isolated from ginger rhizomes (Zingiber officinale), is a novel and selective activator of skeletal and cardiac SR Ca2+-ATPase (EC50=4µM). It is a novel cardiotonic agent with antiplatelet activity. It induces apoptosis in HL-60 cells. Acts as an agonist at vanilloid receptors. Purity: ≥97% (HPLC). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0399