Learn More
GENOVIS INC GINGISKHAN (KGP)
Catalog No. NC0852742
Encompass
GingisKHAN Enzyme GingisKHAN enzyme for digestion of 2 mg of human IgG1, including 5 vials of Reducing Agent. *Unique protease that digests human IgG1 between K223 and T224 producing a homogenous pool of Fab and Fc fragments. The enzyme requires only mild reduced condition for activity and the reaction is performed in the presence of 2 mM Cystein at neutral pH, leaving intact Fab and Fc fragments.
This item is not returnable. View return policy
Catalog No. NC0852742 Supplier GENOVIS INC Supplier No. B0GKH020
GingisKHAN Enzyme GingisKHAN enzyme for digestion of 2 mg of human IgG1, including 5 vials of Reducing Agent. *Unique protease that digests human IgG1 between K223 and T224 producing a homogenous pool of Fab and Fc fragments. The enzyme requires only mild reduced condition for activity and the reaction is performed in the presence of 2 mM Cystein at neutral pH, leaving intact Fab and Fc fragments.