Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginkgolic acid | 20261-38-5 | 99.9% | 320.47 | C20H32O3 | 10 MG
SDP

Catalog No. 5000296669
Encompass_Preferred

Ginkgolic acid (C13:0) is a natural small molecule used in research as an anti-cariogenic agent and a PI3Kδ inhibitor; it also exhibits antibacterial and anti-parasitic activity and can inhibit mast cell degranulation.

  • Chemical formula C20H32O3.
  • Molecular weight 320.47 g·mol-1.
  • CAS number 20261-38-5.
  • Purity 99.92%.
  • Physical form solid suitable for analytical and research use.
  • Available in milligram pack sizes for research (for example, 1 mg, 5 mg, 10 mg).

Catalog No. 50-002-96669 Supplier Medchemexpress LLC Supplier No. HYN007810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ginkgolic acid (C13:0) is a natural small molecule used in research as an anti-cariogenic agent and a PI3Kδ inhibitor; it also exhibits antibacterial and anti-parasitic activity and can inhibit mast cell degranulation.

  • Chemical formula C20H32O3.
  • Molecular weight 320.47 g·mol-1.
  • CAS number 20261-38-5.
  • Purity 99.92%.
  • Physical form solid suitable for analytical and research use.
  • Available in milligram pack sizes for research (for example, 1 mg, 5 mg, 10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.