Learn More
Cayman Chemical GInkgolIc AcId C17 1 10mg
SDP
Catalog No. 502910404
Encompass_Preferred
A phenolic acid with antibacterial and anticancer activities; inhibits EHEC biofilm formation at 5 UG/ml; inhibits JAK2 phosphorylation and STAT3 activation in, and reduces the viability of, U266 cells at 50 µM
Catalog No. 50-291-0404 Supplier Cayman Chemical Supplier No. 3573710 MG
A phenolic acid with antibacterial and anticancer activities; inhibits EHEC biofilm formation at 5 UG/ml; inhibits JAK2 phosphorylation and STAT3 activation in, and reduces the viability of, U266 cells at 50 µM