Learn More
Selleck Chemical LLC Ginkgolide A S2026-25mg
SDP
Catalog No. 503110619
Encompass_Preferred
Ginkgolide A (BN52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist with a Ki of 14 5 M
Catalog No. 50-311-0619 Supplier Selleck Chemical LLC Supplier No. S202625MG
Ginkgolide A (BN52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist with a Ki of 14 5 M