Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginkgolide A from Ginkgo biloba leaves, 90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013216 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3216 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3216 Supplier MilliporeSigma Supelco Supplier No. G402850MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ginkgolide A is a diterpene lactone, which can be extracted from the leaves of Ginkgo biloba tree.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 15291-75-5
Physical Form Neat
Linear Formula C20H24O9
MDL Number MFCD00133365
Type Analytical Standard
Quantity 50 mg
Formula Weight 408.40
Percent Purity ≥90% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.