Learn More
Cayman Chemical GInkgolIde B 10mg
SDP
Catalog No. 502972267
Encompass_Preferred
A potent PAFR antagonist that inhibits platelet aggregation at concentrations from 10-500 μM; demonstrates anticancer, anti-inflammatory, anti-allergic, antioxidant, and neuroprotective effects
Catalog No. 50-297-2267 Supplier Cayman Chemical Supplier No. 1463610 MG
A potent PAFR antagonist that inhibits platelet aggregation at concentrations from 10-500 μM; demonstrates anticancer, anti-inflammatory, anti-allergic, antioxidant, and neuroprotective effects